Learn More
Lonza Walkersville SCBM Stromal Cell Growth Basa
Catalog No. 5000496374
Encompass
SCBM is a Stromal Cell Basal Medium sold separately This can be used with 1 kit of CC-4181 (Stromal Cell Growth Medium SingleQuots Supplements and Growth Factors) The full SCGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3205 Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts SCGM Medium contains 5% FBS Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells Media systems have been specifically developed and optimized for various cell systems
Catalog No. 50-004-96374 Supplier Lonza Walkersville Supplier No. CC3204
Add to Cart
Add to Cart
SCBM is a Stromal Cell Basal Medium sold separately This can be used with 1 kit of CC-4181 (Stromal Cell Growth Medium SingleQuots Supplements and Growth Factors) The full SCGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3205 Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts SCGM Medium contains 5% FBS Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells Media systems have been specifically developed and optimized for various cell systems