Learn More
ATCC SCC-15; Tongue Carcinoma; Human (Homo Sapiens)
Catalog No. 502382988
Encompass
SCC-15 is a cell line that was isolated from the tongue of a 55-year-old, male patient with squamous cell carcinoma. This cell line was deposited by JG Rheinwald and can be used in cancer research.
Catalog No. 50-238-2988 Supplier ATCC Supplier No. CRL1623
SCC-15 is a cell line that was isolated from the tongue of a 55-year-old, male patient with squamous cell carcinoma. This cell line was deposited by JG Rheinwald and can be used in cancer research.