Learn More
ATCC SCC-25; Tongue Carcinoma; Human (Homo sapiens)
Catalog No. 502381840
Encompass
SCC-25 is an epithelial-like cell that was isolated from the tongue of a 70-year-old, male patient with squamous cell carcinoma. This cell line was deposited by JG Rheinwald and can be used in toxicology and cancer research.
Catalog No. 50-238-1840 Supplier ATCC Supplier No. CRL1628
SCC-25 is an epithelial-like cell that was isolated from the tongue of a 70-year-old, male patient with squamous cell carcinoma. This cell line was deposited by JG Rheinwald and can be used in toxicology and cancer research.