Learn More
Aviva Systems Biology Corp SCCPDH Antibody - C-terminal region (ARP53711_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53711_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCCPDH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SCCPDH. Alternate names: CGI-49, FLJ43187, RP11-439E19.2, NET11ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-61103