Learn More
Aviva Systems Biology Corp SCD Antibody - middle region (ARP32797_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32797_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SCD. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SCD. Alternate names: SCD1, FADS5, SCDOS, MSTP008ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rat, Sheep
Catalog No. 50-014-52874