Learn More
Medchemexpress LLC SCD1 inhibitor-1 | 1069094-65-0 | 99.9% | 451.54 | 100 MG
SDP
Catalog No. 5000437444
Encompass_Preferred
SCD1 inhibitor-1 (Compound 48) is an orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1) with an IC50 of 8.8 nM for recombinant human SCD1 enzyme. This compound can be used in the study of diseases such as diabetes, hepatic steatosis, and obesity.
  • Orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1).
  • Improved glucose tolerance and decreased body weight in high-fat mice models.
  • Inhibits SCD1 activity in epididymal adipose tissue and the liver in high-sucrose rat models.
Catalog No. 50-004-37444 Supplier Medchemexpress LLC Supplier No. HY112812100MG
SCD1 inhibitor-1 (Compound 48) is an orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1) with an IC50 of 8.8 nM for recombinant human SCD1 enzyme. This compound can be used in the study of diseases such as diabetes, hepatic steatosis, and obesity.
  • Orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1).
  • Improved glucose tolerance and decreased body weight in high-fat mice models.
  • Inhibits SCD1 activity in epididymal adipose tissue and the liver in high-sucrose rat models.