Learn More
Medchemexpress LLC SCD1 inhibitor-1 | 1069094-65-0 | 99.91% | 451.54 | 50 MG
SDP
Catalog No. 5000435697
Encompass_Preferred
SCD1 inhibitor-1 (Compound 48) is an orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1) with an IC50 of 8.8 nM for recombinant human SCD1 enzyme. It can be utilized in the study of various conditions such as diabetes, hepatic steatosis, and obesity.
  • Orally active.
  • Liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1).
  • Exhibits an IC50 of 8.8 nM for recombinant human SCD1 enzyme.
  • Applicable in the study of diseases including diabetes, hepatic steatosis, and obesity.
  • Improved glucose tolerance in a dose-dependent manner in high-fat mice models.
  • Decreased body weight in a mild dose-dependent manner in high-fat mice models.
  • Inhibited SCD1 inhibitory activity in a dose-dependent manner in epididymal adipose tissue and the liver of high-sucrose rat models.
  • Shows selective tissue distribution, specifically in the liver/eyelid.
Catalog No. 50-004-35697 Supplier Medchemexpress LLC Supplier No. HY11281250MG
SCD1 inhibitor-1 (Compound 48) is an orally active and liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1) with an IC50 of 8.8 nM for recombinant human SCD1 enzyme. It can be utilized in the study of various conditions such as diabetes, hepatic steatosis, and obesity.
  • Orally active.
  • Liver-selective inhibitor of stearoyl-CoA desaturase 1 (SCD1).
  • Exhibits an IC50 of 8.8 nM for recombinant human SCD1 enzyme.
  • Applicable in the study of diseases including diabetes, hepatic steatosis, and obesity.
  • Improved glucose tolerance in a dose-dependent manner in high-fat mice models.
  • Decreased body weight in a mild dose-dependent manner in high-fat mice models.
  • Inhibited SCD1 inhibitory activity in a dose-dependent manner in epididymal adipose tissue and the liver of high-sucrose rat models.
  • Shows selective tissue distribution, specifically in the liver/eyelid.