Learn More
AdipoGen SCD1 ms pAb
SDP
Supplier:  AdipoGen AG25A0031C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse SCD1. Detects a band of ~41kDa by Western Blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Stearoyl-CoA desaturase SCD is a regulatory enzyme in lipogenesis. The enzyme is constitutively expressed in adipose tissue and is induced in liver in response to fat free, high carbohydrate diet. Several SCD isoforms SCD1, -2, and -3 exist in the mouse. Mice with a targeted disruption of the SCD1 isoform had reduced body adiposity, increased insulin sensitivity, and resistance to diet-induced weight gain. The protection from obesity involved increased energy expenditure and increased oxygen consumption. SCD1 deficiency shows activation of lipid oxidation in addition to reduced triglyceride synthesis and storage.
Catalog No. 50-256-3281