Learn More
New England Biolabs, Inc. Sce PUS1 – 5000 pmol
SDP
Catalog No. 502023987
Encompass
Sce Pseudouridine Synthase I (Sce PUS1) is expressed as a 6X His tagged recombinant protein and converts Uridine to Pseudouridine in RNA.  Sce PUS1 has a preference for Uridines in single stranded RNA regions over Uridines in double-stranded RNA. Optimal substrate is an unstructured RNA that is 15 nt long or longer. Sequence-specific pseudouridine modification by Sce PUS1 is an alternative to randomly incorporated modified nucleosides by RNA polymerases.
Catalog No. 50-202-3987 Supplier New England Biolabs, Inc. Supplier No. M0526S
Sce Pseudouridine Synthase I (Sce PUS1) is expressed as a 6X His tagged recombinant protein and converts Uridine to Pseudouridine in RNA.  Sce PUS1 has a preference for Uridines in single stranded RNA regions over Uridines in double-stranded RNA. Optimal substrate is an unstructured RNA that is 15 nt long or longer. Sequence-specific pseudouridine modification by Sce PUS1 is an alternative to randomly incorporated modified nucleosides by RNA polymerases.