Learn More
Cayman Chemical Sceletum trtuosum Mesem 10mg
SDP
Catalog No. 502981590
Encompass_Preferred
An alkaloid with 5-HT reuptake inhibitory activity; selectively binds to the 5-HT transporter (SERT; IC50 = 4.3 UG/ml) over a variety of receptors (IC50s = 148->750 UG/ml); inhibits 5-HT reuptake in vitro (Ki = 1.4 nM); has analgesic effects in the hot plate test at 20 mg/kg without inducing preference in the CPP test
Catalog No. 50-298-1590 Supplier Cayman Chemical Supplier No. 2110010 MG
An alkaloid with 5-HT reuptake inhibitory activity; selectively binds to the 5-HT transporter (SERT; IC50 = 4.3 UG/ml) over a variety of receptors (IC50s = 148->750 UG/ml); inhibits 5-HT reuptake in vitro (Ki = 1.4 nM); has analgesic effects in the hot plate test at 20 mg/kg without inducing preference in the CPP test