Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Scented Paraffin Repellent
SDP

Catalog No. 22445621 Shop All Mopec Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-445-621 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-621 Supplier Mopec™ Supplier No. SP210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Scented Paraffin Repellent prevents wax from sticking to any type surface.

  • Comes in 4 oz. spray bottle
  • Natural and non-toxic
  • Cucumber melon scent

Specifications

Physical Form Liquid
Boiling Point 214°F
Color White
pH 9.7
Quantity 1 Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.