Learn More
MilliporeSigma™ Scepter™ Test Bead Vial
Catalog No. PHCC3BEADS Shop All MilliporeSigma Products
Change view
Click to view available options
Particle Size:
11 μm
Catalog No. Particle Size
PHCC3BEADS 11 μm
1 options
Catalog No. PHCC3BEADS Supplier MilliporeSigma™ Supplier No. PHCC3BEADS
Add to Cart
Add to Cart
Scepter™ test beads can be used to assess the Scepter™ cell counter
  • Scepter™ Test Beads can be used to assess the Scepter™ Cell Counter
  • Compatible with Scepter™ 2.0 and 3.0
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Bead Vial
Particle Size 11 μm
Product Line Scepter
For Use With (Application) Cell Counting