Learn More
ACROBiosystems ACROBiosystems Biotinylated Human SCF / KITLG Protein, Avitag™,His Tag
Supplier:  ACROBiosystems SCFH82E1200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human SCF, Avitag,His Tag (SCF-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 190 (Accession # AAH69797).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 21.7 kDa. The protein migrates as 26-38 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SCF, Avitag,His Tag (Cat. No. SCF-H82E1) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-195-7371