Learn More
ACROBiosystems ACROBiosystems Mouse SCF / KITLG Protein, His Tag
Catalog No. 501956720
Encompass_Preferred
Recombinant Protein;1MG;Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession # NP_038626).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 19-30 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 μg/mL (100 μL/well) can bind Mouse SCF, His Tag (Cat. No. SCF-M5228) with a linear range of 0.05-1 ng/mL (QC tested).
Catalog No. 50-195-6720 Supplier ACROBiosystems Supplier No. SCFM52281MG
Recombinant Protein;1MG;Mouse SCF, His Tag (SCF-M5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Ala 189 (Accession # NP_038626).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 19-30 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 μg/mL (100 μL/well) can bind Mouse SCF, His Tag (Cat. No. SCF-M5228) with a linear range of 0.05-1 ng/mL (QC tested).