Learn More
Enzo Life Sciences Stem cell factor (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX2018040100
Encompass_Preferred
Alternative name: SCF, c-Kit ligand, KL, Steel factor, MGF. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥98% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm). MW: ~18.4kDa. Formulation: Lyophilized from 10mM acetic acid (AcOH) at a concentration of 1mg/ml. Sterile filtered. Source: Produced in E. coli. Non-glycosylated protein, containing 165 amino acids. Long Term Storage: -20°C.
Catalog No. 50-200-8003