Learn More
Medchemexpress LLC SCF Human HEK293 100ug
SDP
Catalog No. 5000197045
Encompass_Preferred
Recombinant human Stem Cell Factor (SCF) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. Supplied lyophilized for research use, this protein supports KIT receptor activation and downstream signaling and is suitable for cell-based assays and biochemical studies. Bioactivity has been validated in TF-1 proliferation assays.
  • High purity (>95%) suitable for sensitive assays.
  • Mammalian expression preserves native glycosylation and activity.
  • C-terminal His tag enables straightforward purification and detection.
  • Lyophilized formulation simplifies storage and transport.
  • Validated bioactivity: ED50 ~1.097 ng/mL in TF-1 cell proliferation assay.
  • Storage stability supported by aliquoting at -20°C to -80°C for long-term use.
Catalog No. 50-001-97045 Supplier Medchemexpress LLC Supplier No. HYP70757100UG
Recombinant human Stem Cell Factor (SCF) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. Supplied lyophilized for research use, this protein supports KIT receptor activation and downstream signaling and is suitable for cell-based assays and biochemical studies. Bioactivity has been validated in TF-1 proliferation assays.
  • High purity (>95%) suitable for sensitive assays.
  • Mammalian expression preserves native glycosylation and activity.
  • C-terminal His tag enables straightforward purification and detection.
  • Lyophilized formulation simplifies storage and transport.
  • Validated bioactivity: ED50 ~1.097 ng/mL in TF-1 cell proliferation assay.
  • Storage stability supported by aliquoting at -20°C to -80°C for long-term use.