Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SCF Human HEK293 1mg
SDP

Catalog No. 5000196909
Encompass_Preferred

Recombinant human stem cell factor extracellular domain produced in HEK293 mammalian cells and purified to >95% purity. Supplied lyophilized and C-terminally tagged with a 6xHis affinity tag, this protein is intended for receptor binding, cell signaling studies, and biochemical assays. Follow storage and reconstitution guidelines to preserve activity.

  • Recombinant extracellular domain (E26-H214) of human stem cell factor.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from phosphate buffer with 150 mM NaCl, pH 7.4.
  • Stable when stored frozen; aliquot after reconstitution for long-term storage.

Catalog No. 50-001-96909 Supplier Medchemexpress LLC Supplier No. HYP707571MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human stem cell factor extracellular domain produced in HEK293 mammalian cells and purified to >95% purity. Supplied lyophilized and C-terminally tagged with a 6xHis affinity tag, this protein is intended for receptor binding, cell signaling studies, and biochemical assays. Follow storage and reconstitution guidelines to preserve activity.

  • Recombinant extracellular domain (E26-H214) of human stem cell factor.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from phosphate buffer with 150 mM NaCl, pH 7.4.
  • Stable when stored frozen; aliquot after reconstitution for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.