Learn More
Medchemexpress LLC SCF Human HEK293 500ug
SDP
Catalog No. 5000197044
Encompass_Preferred
Recombinant human stem cell factor (SCF) produced in HEK293 cells and supplied as a purified, C-terminal 6xHis-tagged protein for use in in vitro studies of KIT receptor signaling. The material is provided in multiple pre-measured quantities and includes handling guidance for storage and reconstitution to preserve activity.
  • Expression in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and store at -80°C for long term.
  • Do not reconstitute below 100 μg/mL; follow recommended handling for activity.
  • Suitable for receptor-ligand binding, signaling, and cell proliferation assays.
Catalog No. 50-001-97044 Supplier Medchemexpress LLC Supplier No. HYP70757500UG
Recombinant human stem cell factor (SCF) produced in HEK293 cells and supplied as a purified, C-terminal 6xHis-tagged protein for use in in vitro studies of KIT receptor signaling. The material is provided in multiple pre-measured quantities and includes handling guidance for storage and reconstitution to preserve activity.
  • Expression in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and store at -80°C for long term.
  • Do not reconstitute below 100 μg/mL; follow recommended handling for activity.
  • Suitable for receptor-ligand binding, signaling, and cell proliferation assays.