Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SCF Mouse HEK293 10ug
SDP

Catalog No. 5000184543
Encompass_Preferred

Recombinant mouse Stem Cell Factor (SCF) produced in HEK293 cells with a C-terminal 6xHis tag, supplied lyophilized for convenient storage and handling. This product is intended for in vitro research applications involving KIT receptor signaling, cell proliferation, survival, and related biological assays, and is supplied with supporting documentation including a datasheet and certificate of analysis.

  • Recombinant protein expressed in HEK293 for mammalian post-translational modifications.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Predicted molecular mass 19.3 kDa and greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, for extended shelf life and easy reconstitution.
  • Stable at -20°C for long-term storage; reconstituted stability at 4°C for one week.
  • Supplied with datasheet and certificate of analysis for quality assurance.

Catalog No. 50-001-84543 Supplier Medchemexpress LLC Supplier No. HYP7055510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Stem Cell Factor (SCF) produced in HEK293 cells with a C-terminal 6xHis tag, supplied lyophilized for convenient storage and handling. This product is intended for in vitro research applications involving KIT receptor signaling, cell proliferation, survival, and related biological assays, and is supplied with supporting documentation including a datasheet and certificate of analysis.

  • Recombinant protein expressed in HEK293 for mammalian post-translational modifications.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Predicted molecular mass 19.3 kDa and greater than 95% purity by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, for extended shelf life and easy reconstitution.
  • Stable at -20°C for long-term storage; reconstituted stability at 4°C for one week.
  • Supplied with datasheet and certificate of analysis for quality assurance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.