Learn More
Enzo Life Sciences Stem cell factor (mouse), (recombinant) (2µg)
SDP
Supplier:  Enzo Life Sciences ALX2018050002
Encompass_Preferred
Alternative name: SCF, c-Kit ligand, KL, Steel factor, MGF. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (reducing and non-reducing SDS-PAGE). MW: ~18.3kDa. Appearance: White lyophilized (freeze-dried) powder. Formulation: Lyophilized from a sterile aqueous solution containing 20mM sodium phospate, pH 6.5. Source: Produced in E. coli. Non-glycosylated protein, containing 164 amino acids. Long Term Storage: -20°C. Use/Stability: It is recommended that a carrier protein (0.1% HSA or BSA) is added for long term storage. Handling: Centrifuge the vial before opening the cap. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8004