Learn More
Medchemexpress LLC HY-P7064 10ug Medchemexpress, SCF (P.pastoris-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934307
Encompass_Preferred
Medchemexpress, HY-P7064 10ug SCF (P.pastoris-expressed), Mouse CAS: Recombinant Mouse Stem Cell Factor (P.pastoris-expressed) is the ligand for the receptor-type protein-tyrosine kinase KIT. It plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4307 Supplier Medchemexpress LLC Supplier No. HYP706410UG
Medchemexpress, HY-P7064 10ug SCF (P.pastoris-expressed), Mouse CAS: Recombinant Mouse Stem Cell Factor (P.pastoris-expressed) is the ligand for the receptor-type protein-tyrosine kinase KIT. It plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.