Learn More
Medchemexpress LLC HY-P7056 10ug Medchemexpress, SCF (P.pastoris-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP705610UG
Encompass_Preferred
Medchemexpress, HY-P7056 10ug SCF (P.pastoris-expressed), Human CAS: Recombinant Human Stem Cell Factor (P.pastoris-expressed), the c-kit ligand, is a two disulfide bridge-containing cytokine in the regulation of the development and function of hematopoietic cell lineages and other cells such as mast cells, germ cells, and melanocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4301