Learn More
Sino Biological Recombinant Mouse SCF Protein (His Tag), Amine-Labeled Biotinylated
Catalog No. 502116218
Encompass_Preferred
A DNA sequence encoding the Mouse Kitlg (P20826-1) (Met1-Ala189) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6218 Supplier Sino Biological Supplier No. 50487M08BB100
A DNA sequence encoding the Mouse Kitlg (P20826-1) (Met1-Ala189) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.