Learn More
Medchemexpress LLC SCF Rat HEK293 100ug | 100ug
SDP
Catalog No. 5000196597
Encompass_Preferred
SCF Protein Rat (HEK293) is a hematopoietic cytokine triggered by binding to its ligand c-kit and plays an important role in hematopoiesis spermatogenesis and melanogenesis
Catalog No. 50-001-96597 Supplier Medchemexpress LLC Supplier No. HYP7107A100UG
SCF Protein Rat (HEK293) is a hematopoietic cytokine triggered by binding to its ligand c-kit and plays an important role in hematopoiesis spermatogenesis and melanogenesis