Learn More
Medchemexpress LLC SCF Rat HEK293 Hi 10ug
SDP
Catalog No. 5000196491
Encompass_Preferred
Recombinant rat stem cell factor expressed in HEK293 cells, provided as a lyophilized protein for research use. SCF binds the KIT receptor and regulates hematopoiesis, cell survival, proliferation, and differentiation, making it suitable for cell signaling studies, stem/progenitor cell expansion, and functional assays.
  • High purity suitable for cell-based assays.
  • Supports hematopoietic stem and progenitor cell expansion.
  • Promotes cell survival, proliferation, and differentiation in vitro.
  • Validated in TF-1 cell proliferation assays with low ED50.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Expressed in mammalian cells to preserve post-translational modifications.
Catalog No. 50-001-96491 Supplier Medchemexpress LLC Supplier No. HYP7113010UG
Recombinant rat stem cell factor expressed in HEK293 cells, provided as a lyophilized protein for research use. SCF binds the KIT receptor and regulates hematopoiesis, cell survival, proliferation, and differentiation, making it suitable for cell signaling studies, stem/progenitor cell expansion, and functional assays.
  • High purity suitable for cell-based assays.
  • Supports hematopoietic stem and progenitor cell expansion.
  • Promotes cell survival, proliferation, and differentiation in vitro.
  • Validated in TF-1 cell proliferation assays with low ED50.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Expressed in mammalian cells to preserve post-translational modifications.