Learn More
Medchemexpress LLC SCF Rat HEK293 Hi 50ug
SDP
Catalog No. 5000196515
Encompass_Preferred
Recombinant rat stem cell factor (SCF) produced in HEK293 mammalian cells with a C-terminal His tag, supplied as a lyophilized powder for research use. The protein is formulated for stability and is suitable for biochemical and cell-based assays involving KIT receptor signaling and hematopoiesis.
  • Expressed in HEK293 mammalian cells to support native folding and post-translational modifications.
  • C-terminal His tag to facilitate detection and purification.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.
Catalog No. 50-001-96515 Supplier Medchemexpress LLC Supplier No. HYP7113050UG
Recombinant rat stem cell factor (SCF) produced in HEK293 mammalian cells with a C-terminal His tag, supplied as a lyophilized powder for research use. The protein is formulated for stability and is suitable for biochemical and cell-based assays involving KIT receptor signaling and hematopoiesis.
  • Expressed in HEK293 mammalian cells to support native folding and post-translational modifications.
  • C-terminal His tag to facilitate detection and purification.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.