Learn More
Aviva Systems Biology Corp SCFD1 Antibody - N-terminal region (ARP55851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCFD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SCFD1. Alternate names: C14orf163, KIAA0917, RA410, SLY1, STXBP1L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61635