Learn More
Aviva Systems Biology Corp Scg2 Antibody - middle region (ARP45474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Scg2. It was validated on Western Blot. . Alternate names: ChcgApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-203