Learn More
Aviva Systems Biology Corp SCG2 Antibody - N-terminal region (ARP45473_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45473_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SCG2. Alternate names: CHGC, SN, SgIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-202