Learn More
Aviva Systems Biology Corp SCG3 Antibody - C-terminal region (ARP59444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCG3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SCG3. Alternate names: FLJ90833, SGIIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-699