Learn More
Aviva Systems Biology Corp SCG5 Antibody - N-terminal region (ARP63847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCG5. It was validated on Western Blot. . Alternate names: 7B2, P7B2, SGNE1, SgVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-099