Learn More
Lonza Walkersville SCGM Stromal Cell Growth Medium BulletKit
Catalog No. NC9533545
Encompass
Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts. SCGMmedium contains 5% FBS.Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x Bottle Basal Medium 500 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 25.00 mL
Catalog No. NC9533545 Supplier Lonza Walkersville Supplier No. CC3205
Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts. SCGMmedium contains 5% FBS.Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x Bottle Basal Medium 500 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 25.00 mL