Learn More
Lonza Walkersville SCGM Stromal Cell Growth Med
Catalog No. 5000496400
Encompass
SCGM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the SCGM Stromal Cell Growth Medium BulletKit (Catalog Number CC-3205) The full SCGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3205 Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts SCGM Medium contains 5% FBS Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells Media systems have been specifically developed and optimized for various cell systems
Catalog No. 50-004-96400 Supplier Lonza Walkersville Supplier No. CC4181
SCGM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the SCGM Stromal Cell Growth Medium BulletKit (Catalog Number CC-3205) The full SCGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3205 Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts SCGM Medium contains 5% FBS Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells Media systems have been specifically developed and optimized for various cell systems