Learn More
Aviva Systems Biology Corp SCGN Antibody - middle region (ARP42333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCGN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCGN. Alternate names: CALBL, DJ501N12.8, SECRET, SEGN, setaginApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58188