Learn More
Medchemexpress LLC Sch-1473759 hydrochloride | 1094067-13-6 | MFCD18251447 | 99.2% | 463.00 g/mol | C20H27ClN8OS | 100 MG
SDP
Catalog No. 5000244044
Encompass_Preferred
SCH-1473759 hydrochloride is a research-grade aurora kinase inhibitor supplied as the hydrochloride salt. It exhibits low-nanomolar potency against Aurora A and B kinases and is provided as a high-purity solid for in vitro and preclinical research applications.
  • Selective aurora A and B inhibition (IC50 = 4 nM and 13 nM).
  • High purity suitable for research use.
  • Provided as a solid, white to yellow in appearance.
  • Hydrochloride form improves solubility for assay preparation.
  • Available in multiple pack sizes and a DMSO solution for convenience.
  • Intended for in vitro kinase assays and cell-based studies.
Catalog No. 50-002-44044 Supplier Medchemexpress LLC Supplier No. HY10483100MG
SCH-1473759 hydrochloride is a research-grade aurora kinase inhibitor supplied as the hydrochloride salt. It exhibits low-nanomolar potency against Aurora A and B kinases and is provided as a high-purity solid for in vitro and preclinical research applications.
  • Selective aurora A and B inhibition (IC50 = 4 nM and 13 nM).
  • High purity suitable for research use.
  • Provided as a solid, white to yellow in appearance.
  • Hydrochloride form improves solubility for assay preparation.
  • Available in multiple pack sizes and a DMSO solution for convenience.
  • Intended for in vitro kinase assays and cell-based studies.