Learn More
Abcam SCH-202676, GPCR allosteric modulator, 50MG
Catalog No. 502705604
Encompass_Preferred
MW 348.3 Da, Purity >98%. Non-selective GPCR allosteric modulator. Inhibits adenosine, opioid, muscarinic, adrenergic and dopaminergic receptors (IC₅₀ values are 0.1-1.8 μM). Inhibits agonist and antagonist binding to GPCRs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5604 Supplier Abcam Supplier No. AB14457950MG
MW 348.3 Da, Purity >98%. Non-selective GPCR allosteric modulator. Inhibits adenosine, opioid, muscarinic, adrenergic and dopaminergic receptors (IC₅₀ values are 0.1-1.8 μM). Inhibits agonist and antagonist binding to GPCRs.
The product is subject to the following: Abcam Restricted Use Statement