Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SCH-202676 hydrobromide | 265980-25-4 | 96.8% | 348.26 g·mol⁻¹ | C15H14BrN3S | 5 MG
SDP

Catalog No. 5000263429
Encompass_Preferred

SCH-202676 hydrobromide is a small-molecule allosteric modulator of G protein-coupled receptors and adenosine receptors with reported antiviral activity, including inhibition of 3CLpro. The compound is supplied for research use in solid and solution forms and is intended for biochemical and pharmacological studies.

  • Allosteric modulator of GPCRs and adenosine receptors.
  • Reported antiviral activity; inhibits 3CLpro with low-micromolar potency.
  • Available as solid and 10 mM solution in DMSO.
  • High purity (96.81%).
  • Molecular formula C15H14BrN3S; molecular weight 348.26 g·mol⁻¹.
  • Store at -20°C long term; in solvent store at -80°C for extended stability.

Catalog No. 50-002-63429 Supplier Medchemexpress LLC Supplier No. HY1100125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SCH-202676 hydrobromide is a small-molecule allosteric modulator of G protein-coupled receptors and adenosine receptors with reported antiviral activity, including inhibition of 3CLpro. The compound is supplied for research use in solid and solution forms and is intended for biochemical and pharmacological studies.

  • Allosteric modulator of GPCRs and adenosine receptors.
  • Reported antiviral activity; inhibits 3CLpro with low-micromolar potency.
  • Available as solid and 10 mM solution in DMSO.
  • High purity (96.81%).
  • Molecular formula C15H14BrN3S; molecular weight 348.26 g·mol⁻¹.
  • Store at -20°C long term; in solvent store at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.