Learn More
Medchemexpress LLC SCH-202676 hydrobromide | 265980-25-4 | 96.81% | 348.26 | 1 MG
SDP
Catalog No. 5000345467
Encompass_Preferred
SCH-202676 hydrobromide is an allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR). It exhibits antiviral activity and inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
  • Allosteric modulator of G protein-coupled receptors (GPCRs)
  • Allosteric modulator of adenosine receptor (AR)
  • Antiviral activity
  • Inhibits 3CLpro in a time-dependent manner
  • IC50 value of 0.655 μM against 3CLpro
Catalog No. 50-003-45467 Supplier Medchemexpress LLC Supplier No. HY11001250MG
SCH-202676 hydrobromide is an allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR). It exhibits antiviral activity and inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
  • Allosteric modulator of G protein-coupled receptors (GPCRs)
  • Allosteric modulator of adenosine receptor (AR)
  • Antiviral activity
  • Inhibits 3CLpro in a time-dependent manner
  • IC50 value of 0.655 μM against 3CLpro