Learn More
Medchemexpress LLC SCH-202676 hydrobromide | 265980-25-4 | 96.81% | 348.26 | 100 MG
SDP
Catalog No. 5000343475
Encompass_Preferred
SCH-202676 hydrobromide is an allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR). It exhibits antiviral activity and inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
  • Allosteric modulator of G protein-coupled receptors and adenosine receptor.
  • Exhibits antiviral activity.
  • Inhibits 3CLpro in a time-dependent manner.
  • IC50 value of 0.655 μM.
Catalog No. 50-003-43475 Supplier Medchemexpress LLC Supplier No. HY11001210MM/1ML
SCH-202676 hydrobromide is an allosteric modulator of G protein-coupled receptors (GPCRs) and adenosine receptor (AR). It exhibits antiviral activity and inhibits 3CLpro in a time-dependent manner with an IC50 value of 0.655 μM.
  • Allosteric modulator of G protein-coupled receptors and adenosine receptor.
  • Exhibits antiviral activity.
  • Inhibits 3CLpro in a time-dependent manner.
  • IC50 value of 0.655 μM.