Learn More
Cayman Chemical SCH 202676 (hydrobromide), Purity: ≥95%, CAS: 265980-25-4, 10 mg
SDP
Catalog No. 502965739
Encompass_Preferred
A reversible inhibitor of both agonist and antagonist binding to diverse GPCRs; blocks the binding of radiolabeled ligands to human opioid, adrenergic, muscarinic, dopaminergic, adenosine, and purinergic receptors with IC50 values of 0.1 to 1.8 µM
Catalog No. 50-296-5739 Supplier Cayman Chemical Supplier No. 1694510 MG
A reversible inhibitor of both agonist and antagonist binding to diverse GPCRs; blocks the binding of radiolabeled ligands to human opioid, adrenergic, muscarinic, dopaminergic, adenosine, and purinergic receptors with IC50 values of 0.1 to 1.8 µM