Learn More
TARGETMOL CHEMICALS INC SCH 221510 5MG
SDP
Catalog No. 502580878
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SCH 221510 is an orally available selective and potent NOP agonist with an EC50 of 12 nM and a Ki of 0.3 nM.SCH 221510 has anxiolytic activity and can be used to study neurological disorders. purity: 99%
Catalog No. 50-258-0878 Supplier TARGETMOL CHEMICALS INC Supplier No. T233355MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SCH 221510 is an orally available selective and potent NOP agonist with an EC50 of 12 nM and a Ki of 0.3 nM.SCH 221510 has anxiolytic activity and can be used to study neurological disorders. purity: 99%