Learn More
Medchemexpress LLC SCH-23390 hydrochloride | 125941-87-9 | 99.8% | 324.24 | 1 ML
SDP
Catalog No. 5000401941
Encompass_Preferred
SCH-23390 hydrochloride is a potent and selective dopamine D1-like receptor antagonist with Kis of 0.2 nM and 0.3 nM for the D1 and D5 receptor, respectively. It is also a potent and high efficacy human 5-HT2C receptor agonist with a Ki of 9.3 nM, and binds with high affinity to 5-HT2 and 5-HT1C receptors. It inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels with an IC50 of 268 nM. For research use only.
  • Potent and selective dopamine D1-like receptor antagonist.
  • Potent and high efficacy human 5-HT2C receptor agonist.
  • Binds with high affinity to 5-HT2 and 5-HT1C receptors.
  • Inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels.
  • Used in studies of neurological disorders like psychosis and Parkinson's disease.
  • Used in topographical determination of brain D1 receptors.
Catalog No. 50-004-01941 Supplier Medchemexpress LLC Supplier No. HY19545A10MM/1ML
SCH-23390 hydrochloride is a potent and selective dopamine D1-like receptor antagonist with Kis of 0.2 nM and 0.3 nM for the D1 and D5 receptor, respectively. It is also a potent and high efficacy human 5-HT2C receptor agonist with a Ki of 9.3 nM, and binds with high affinity to 5-HT2 and 5-HT1C receptors. It inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels with an IC50 of 268 nM. For research use only.
  • Potent and selective dopamine D1-like receptor antagonist.
  • Potent and high efficacy human 5-HT2C receptor agonist.
  • Binds with high affinity to 5-HT2 and 5-HT1C receptors.
  • Inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels.
  • Used in studies of neurological disorders like psychosis and Parkinson's disease.
  • Used in topographical determination of brain D1 receptors.