Learn More
TARGETMOL CHEMICALS INC SCH-23390 HYDROCHLORIDE 25MG
SDP
Catalog No. 502579896
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH-23390 hydrochloride (R-(+)-SCH23390 hydrochloride) is an effective dopamine receptor antagonist and for the D1(Ki=0.2 nM) and D5(Ki=0.3 nM). purity: 99%
Catalog No. 50-257-9896 Supplier TARGETMOL CHEMICALS INC Supplier No. T436925MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH-23390 hydrochloride (R-(+)-SCH23390 hydrochloride) is an effective dopamine receptor antagonist and for the D1(Ki=0.2 nM) and D5(Ki=0.3 nM). purity: 99%