Learn More
Medchemexpress LLC SCH-23390 hydrochloride (R-(+)-SCH-23390 hydrochloride) | 125941-87-9 | 99.8% | 324.24 | 1 MG
SDP
Catalog No. 5000400725
Encompass_Preferred
SCH-23390 hydrochloride is a potent and selective dopamine D1-like receptor antagonist, also acting as a potent human 5-HT2C receptor agonist. It binds with high affinity to various serotonin receptors and inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels. This compound can reverse inhibitory effects on NLRP3 expression and inflammasome pathways in BV-2 cells, and has been used in studies of neurological disorders like psychosis and Parkinson's disease.
  • Potent and selective dopamine D1-like receptor antagonist
  • Potent and high efficacy human 5-HT2C receptor agonist
  • Binds with high affinity to 5-HT2 and 5-HT1C receptors
  • Inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels
  • Used in studies of neurological disorders like psychosis and Parkinson's disease
Catalog No. 50-004-00725 Supplier Medchemexpress LLC Supplier No. HY19545A1MG
SCH-23390 hydrochloride is a potent and selective dopamine D1-like receptor antagonist, also acting as a potent human 5-HT2C receptor agonist. It binds with high affinity to various serotonin receptors and inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels. This compound can reverse inhibitory effects on NLRP3 expression and inflammasome pathways in BV-2 cells, and has been used in studies of neurological disorders like psychosis and Parkinson's disease.
  • Potent and selective dopamine D1-like receptor antagonist
  • Potent and high efficacy human 5-HT2C receptor agonist
  • Binds with high affinity to 5-HT2 and 5-HT1C receptors
  • Inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels
  • Used in studies of neurological disorders like psychosis and Parkinson's disease