Learn More
Cayman Chemical SCH 23390 (hydrochloride), Purity: ≥99%, CAS: 125941-87-9, 50 mg
SDP
Catalog No. 502972241
Encompass_Preferred
A selective antagonist of the dopamine D1-like receptor subtypes D1 and D5 (Kis = 0.2 and 0.3 nM, respectively); useful for studying the role of the dopamine system in normal brain function and neurological disorders
Catalog No. 50-297-2241 Supplier Cayman Chemical Supplier No. 1563150 MG
A selective antagonist of the dopamine D1-like receptor subtypes D1 and D5 (Kis = 0.2 and 0.3 nM, respectively); useful for studying the role of the dopamine system in normal brain function and neurological disorders