Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCH 28080, Tocris Bioscience™

Catalog No. 169010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-901-0 10 mg
16-905-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-901-0 Supplier R&D Systems Supplier No. 1690/10
This item is not returnable. View return policy

H+,K+-ATPase inhibitor

Potent inhibitor of H+,K+-ATPase (IC50 = 20 nM); binds to the K+ recognition site and is competitive with respect to K+. Inhibits gastric acid secretion in vitro and in vivo.

Chemical Identifiers

CAS 76081-98-6
Molecular Formula C17H15N3O
Molecular Weight (g/mol) 277.33
MDL Number MFCD00834620
InChI Key PYKJFEPAUKAXNN-UHFFFAOYSA-N
Synonym schering compound 28080, 2-methyl-8-phenylmethoxy imidazo 1,2-a pyridine-3-acetonitrile, chembl47529, 2-methyl-8-phenylmethoxy imidazo-1-2-a pyrine-3-acetonitrile, 2-8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, imidazo 1,2-a pyridine-3-acetonitrile, 2-methyl-8-phenylmethoxy, tocris-1690, 8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, lopac-s-4443
PubChem CID 108137
IUPAC Name 2-[8-(benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl]acetonitrile
SMILES CC1=C(CC#N)N2C=CC=C(OCC3=CC=CC=C3)C2=N1

Specifications

Quantity 10 mg
Formula Weight 277.33
Percent Purity >99%
Chemical Name or Material SCH 28080
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.