Learn More
Cayman Chemical SCH 38519 2.5mg
SDP
Catalog No. 502938891
Encompass_Preferred
A fungal metabolite; inhibits thrombin-induced aggregation, as well as inhibits serotonin secretion in human platelets (IC50s = 68 and 61 UG/ml, respectively); inhibits the growth of various Gram-negative and Gram-positive bacteria (mean MICs = 122.1 and 0.92 UG/ml, respectively)
Catalog No. 50-293-8891 Supplier Cayman Chemical Supplier No. 280922.5 MG
A fungal metabolite; inhibits thrombin-induced aggregation, as well as inhibits serotonin secretion in human platelets (IC50s = 68 and 61 UG/ml, respectively); inhibits the growth of various Gram-negative and Gram-positive bacteria (mean MICs = 122.1 and 0.92 UG/ml, respectively)