Learn More
Apexbio Technology LLC SCH 527123 473727-83-2 200mg
SDP
Catalog No. 5000064344
Encompass_Preferred
SCH 527123 (CAS 473727-83-2) is a selective antagonist of chemokine receptor 2 (CXCR2) It inhibits CXCR2-mediated signaling pathways by reducing phosphorylation events within the NF- B MAPK and AKT signaling cascades In colorectal cancer cell lines SCH 527123 suppresses cell growth in a dose-dependent manner with reported IC50 values ranging from approximately 18 to 40 mol/L after a 72-hour treatment Elevated IL-8 expression in cell lines such as HCT116 and Caco2 correlates with decreased sensitivity to SCH 527123 This compound is utilized in research investigating CXCR2 signaling in inflammation and tumor growth
Catalog No. 50-000-64344 Supplier Apexbio Technology LLC Supplier No. A3802200
SCH 527123 (CAS 473727-83-2) is a selective antagonist of chemokine receptor 2 (CXCR2) It inhibits CXCR2-mediated signaling pathways by reducing phosphorylation events within the NF- B MAPK and AKT signaling cascades In colorectal cancer cell lines SCH 527123 suppresses cell growth in a dose-dependent manner with reported IC50 values ranging from approximately 18 to 40 mol/L after a 72-hour treatment Elevated IL-8 expression in cell lines such as HCT116 and Caco2 correlates with decreased sensitivity to SCH 527123 This compound is utilized in research investigating CXCR2 signaling in inflammation and tumor growth