Learn More
Apexbio Technology LLC SCH 527123, 5mg. Cas: 473727-83-2 MFCD: MFCD16628072. CXCR1 and CXCR2 receptors antagonist
SDP
Supplier:  Apexbio Technology LLC A38025
Encompass_Preferred
SCH-527123 is a novel, selective CXC chemokine receptor 2 (CXCR2) antagonist.SCH-527123 was able to suppress CXCR2-mediated signal transduction as shown through decreased phosphorylation of the NF-B/mitogen-activated protein kinase (MAPK)/AKT pathway [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4842