Learn More
Medchemexpress LLC HY-10017 10mg Medchemexpress, SCH 546738 CAS:906805-42-3 Purity:>98%
SDP
Catalog No. 502028610
Encompass_Preferred
Medchemexpress, HY-10017 10mg SCH 546738 CAS:906805-42-3 SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8610 Supplier Medchemexpress LLC Supplier No. HY1001710MG
Medchemexpress, HY-10017 10mg SCH 546738 CAS:906805-42-3 SCH 546738 is a potent, orally active and non-competitive CXCR3 antagonist, the affinity constant (Ki) of SCH 546738 binding to human CXCR3 receptor is determined to be 0.4 nM in multiple experiments. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.